Learn More
Medchemexpress LLC H-8 dihydrochloride | 113276-94-1 | 99.2% | 338.25 | 10 MG

Supplier: Medchemexpress LLC HY11246510MG
H-8 (dihydrochloride) is a cell-permeable, reversible and ATP-competitive PKA inhibitor. This product is for research use only and not sold to patients.
- Cell-permeable, reversible, and ATP-competitive PKA inhibitor
- For research use only
- Purity: 99.19%
- Molecular weight: 338.25
- Formula: C12H17Cl2N3O2S
- Appearance: Solid
- Color: White to off-white
- Shipping at room temperature (continental US)
- Store at -20°C, sealed, away from moisture
- Soluble in DMSO at 20.83 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.