Learn More
Selleck Chemical LLC H 89 2HCl 10mM/1mL 130964-39-5
SDP
Supplier:  Selleck Chemical LLC S158210MM/1ML
Encompass_Preferred
H 89 2HCl is a potent PKA inhibitor with Ki of 48 nM in a cell-free assay, 10-fold selective for PKA than PKG,500-fold greater selectivity than PKC, MLCK, calmodulin kinase II and casein kinase I/II. H 89 2HCl induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1405