Learn More
Apexbio Technology LLC H 89 2HCl 130964-39-5 200mg
SDP
Supplier:  Apexbio Technology LLC B2190200
Encompass_Preferred
H 89 2HCl (CAS 130964-39-5) is a small-molecule inhibitor targeting cyclic AMP-dependent protein kinase (PKA) It exhibits a Ki of approximately 48 nM toward PKA and demonstrates substantial selectivity compared to related kinases such as PKG (about 10-fold) PKC MLCK calmodulin kinase II and casein kinases (approximately 500-fold) In PC12D cells H 89 at micromolar concentrations suppresses forskolin-induced phosphorylation events and neurite outgrowth without altering intracellular cAMP levels Due to its selective inhibitory profile H 89 is widely utilized in cell signaling studies examining cAMP-dependent pathways
Catalog No. 50-000-61571