Learn More
AdipoGen H-89 . dihydrochloride (25 mg)
SDP
Supplier:  AdipoGen AGCR10002M025
Encompass_Preferred
Chemical. CAS: 130964-39-5 | 127243-85-0 (free base). Formula: C20H20BrN3O2S . 2HCl. MW: 446.4 . 72.9. Cell permeable potent and selective cAMP- and cGMP-dependent protein kinase (PKA and PKG) inhibitor. Protein kinase Cµ (PKCµ) inhibitor. Ca2+/calmodulin-dependent protein kinase II inhibitor. Casein kinase I inhibitor. Myosin light chain kinase (MLCK) inhibitor. Apoptosis enhancer. Tool to study protein crystal structure-inhibitor interactions. Rho kinase inhibitor. Cell proliferation inhibitor. Review.
Catalog No. 50-205-3747