Learn More
Medchemexpress LLC H-9 dihydrochloride | 116700-36-8 | MFCD00069286 | 99.0% | 324.23 g·mol⁻¹ | C11H15Cl2N3O2S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10092350MG
Encompass_Preferred
H-9 dihydrochloride is a cell-permeable small-molecule inhibitor of protein kinase A (PKA) supplied as a research reagent for in vitro biochemical and cellular studies. It is used to probe PKA-dependent signaling pathways and phosphorylation-dependent processes, and is provided with high reported purity suitable for assay applications.
  • Potent PKA inhibition for signaling studies.
  • High reported purity suitable for biochemical assays.
  • Available in small quantities for dose-response and optimization.
  • Stable solid form for convenient storage and handling.
  • Applicable to in vitro kinase assays and cellular experiments.
Catalog No. 50-002-12388