Learn More
Abcam H 9 dihydrochloride, Protein kinase inhibitor, 50MG
Supplier:  Abcam AB14613250MG
Encompass_Preferred
MW 324.2 Da, Purity >99%. Protein kinase inhibitor (Ki values are 0.9 (PKG), 1.9 (PKA), 18 (PKC), 60 (CaMK II), 110 (casein kinase I) and >300 μM (casein kinase II)) similar to H8 dihydrochloride (ab142309) and H7 (ab142308). Inhibits CTL-mediated lysis in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4939