Learn More
Cayman Chemical H-9hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 133125 MG
Encompass_Preferred
A potent competitive inhibitor of PKC, PKG, and PKA with Ki values of 18, 0.87, and 1.9 µM, repectively
Catalog No. 50-295-4751