Learn More
eMolecules​ H-Aib-OEt hydrochloride | Combi-Blocks | 17288-15-2 | MFCD06809830 | 167.630 | C6H14ClNO2 | 98.000 | Cl.CCOC(=O)C(C)(C)N | 25g | 228815167
Supplier:  eMolecules​ 228815167
Encompass_Preferred
H-Aib-OEt hydrochloride | Combi-Blocks | 17288-15-2 | MFCD06809830 | 167.630 | C6H14ClNO2 | 98.000 | Cl.CCOC(=O)C(C)(C)N | 25g | 228815167
Catalog No. NC2219947