Learn More
Medchemexpress LLC H-Ala-d-Ala-OH | 3695-80-5 | MFCD00066040 | 98.2% | 160.17 g/mol | C6H12N2O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW25071525MG
Encompass_Preferred
H-Ala-d-Ala-OH is a dipeptide (alanyl-D-alanine) biochemical reagent for laboratory research use. It is supplied as a solid with a reported purity of 98.21% and a molecular weight of 160.17 g/mol. The compound dissolves in water and is used in biochemical assays and peptide research; follow recommended storage conditions and protect from light.
Catalog No. 50-002-80609