Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Ala-Hyp-OH | 76400-25-4 | 99.4% | 202.21 | 10 MM 1 ML
SDP

Supplier:  Medchemexpress LLC HYP533810MM/1ML

Encompass_Preferred

H-Ala-Hyp-OH (L-Alanyl-(4R)-4-hydroxy-L-proline) is a biologically active peptide supplied as a 10 mM solution in 1 mL of DMSO, primarily intended for research use. It is a white to off-white solid with a high purity of 99.36%.

  • Biologically active peptide
  • Purity of 99.36%
  • Molecular weight of 202.21
  • Supplied as a 10 mM solution in DMSO
  • White to off-white solid appearance

Catalog No. 50-003-99627


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.