Learn More
Medchemexpress LLC H-Arg-4MβNA | 60285-94-1 | 99.6% | 329.40 | 10 MM x 1 ML
SDP
Supplier:  Medchemexpress LLC HY11268810MM/1ML
Encompass_Preferred
H-Arg-4MβNA is a substrate for cathepsin H, used for the detection of enzyme activity in gel electrophoresis.
  • Substrate for cathepsin H
  • Used for the detection of enzyme activity in gel electrophoresis
  • Appearance: Solid
  • Color: White to off-white
  • For research use only.
Catalog No. 50-004-37450