Learn More
Bachem H-Arg-Arg-OH
SDP
Supplier:  Bachem 4000091.1000
Encompass_Preferred
H-Arg-Arg-OH Acetate, 15483-27-9, C12H26N8O3, Mr 330.39, 1g. Arginine-containing dipeptides as RR and RF are used by nitric oxide synthase as substrates in place of Arg.
Catalog No. 50-259-273