Learn More
Medchemexpress LLC H-Asp-OMe | 17812-32-7 | 95.0% | 147.13 g·mol⁻¹ | C5H9NO4 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01749910G
Encompass_Preferred
H-Asp-OMe is the methyl ester derivative of aspartic acid provided as a white to off-white solid for research use. It serves as an amino acid derivative and building block in peptide synthesis and related chemical biology studies, with typical purity suitable for many laboratory applications.
  • Methyl ester of aspartic acid useful as a building block in peptide synthesis.
  • White to off-white solid, convenient for handling and weighing.
  • 95.0% purity suitable for many research applications.
  • Available in multiple pack sizes suitable for small- and mid-scale work.
  • Powder storage stability: -20°C for 3 years or 4°C for 2 years.
  • In solvent storage recommendation: -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-002-99980