Learn More
Medchemexpress LLC H-Asp(OtBu)-OMe HCl | 2673-19-0 | MFCD00153427 | 98.0% | 239.70 g·mol⁻1 | C9H18ClNO4 | 10 G

Supplier: Medchemexpress LLC HYW01848910G
H-Asp(OtBu)-OMe·HCl is an aspartic acid derivative supplied as the methyl ester hydrochloride with the side chain protected as tert-butyl. It is used as a protected amino acid building block in peptide synthesis and organic chemistry to enable selective coupling and controlled deprotection during multi-step assemblies.
- Side-chain tert-butyl protection enables selective deprotection
- Methyl ester HCl salt for improved stability and handling
- High purity suitable for synthesis applications (98.0%)
- Solid form, easy to weigh and transfer
- Store sealed at 4°C, away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.