Learn More
BIOHIPPO h BID shRNA Set RFP BSD
SDP
Supplier:  BIOHIPPO BHV194001472
Encompass
h BID shRNA Set (RFP BSD) shRNA lentiviral vector expressing RFP with Blasticidin selection for studies in Human Each vial provides a nominal titer of 5x10E6 (sh-mix) 5x10E6 (scr-mix) transducing units TU Validated human BID shRNA Lentivirus High-titer shRNA lentiviral particles specific for human BH3 interacting domain death (BID) The shRNA has been validated to meet or exceed 70% BID knockdown efficiency using a specific fluorescence-based method that is more rapid and reliable than qPCR The shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs
Catalog No. 50-003-30960