Learn More
Medchemexpress LLC H-Cyclopropyl-Gly-OH | 49606-99-7 | 115.13 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00575910G
Encompass_Preferred
H-Cyclopropyl-Gly-OH is a glycine derivative, a type of amino acid derivative. These substances are recognized as beneficial ergogenic dietary supplements, known to influence anabolic hormone secretion, supply fuel during exercise, improve mental performance during stress, and prevent exercise-induced muscle damage.
  • Glycine derivative
  • Amino acid derivative
  • Functions as an ergogenic supplement
  • Aids in anabolic hormone secretion
  • Provides fuel during exercise
  • Supports mental performance under stress
  • Helps prevent exercise-induced muscle damage
  • White to off-white solid appearance
  • Soluble in water (10 mg/mL with sonication)
Catalog No. 50-004-48591