Learn More
Selleck Chemical LLC H-Cys
SDP
Supplier:  Selleck Chemical LLC S552225MG
Encompass_Preferred
H-Cys(Trt)-OH (L-Cys(Trt)-OH H-L-Cys(Trt)-OH S-Tritylcysteine S-Trityl-L-cysteine) is a specific inhibitor of Eg5 that inhibits Eg5-driven microtubule sliding velocity in a reversible fashion with an IC50 of 500 nM
Catalog No. 50-000-03492