Learn More
Medchemexpress LLC H-Cys(Bzl)-OH | 3054-01-1 | MFCD00002613 | 99.6% | 211.28 g/mol | C10H13NO2S | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01432910G
Encompass_Preferred
H-Cys(Bzl)-OH is a benzyl-protected cysteine derivative used as a building block in peptide synthesis and other organic chemistry applications. Supplied as a solid with high purity, this reagent is intended for research use and is provided with analytical documentation to support compound identity and quality.
  • Benzyl protection on the thiol for selective coupling.
  • High purity suitable for synthesis and analysis.
  • Supplied as a solid, white to off-white, for easy handling.
  • Available in lab-scale package sizes, including 10 g.
  • Analytical data available (COA, HNMR, LCMS).
  • Recommended sealed storage to minimize moisture exposure.
Catalog No. 50-003-04097