Learn More
Bachem H-Cys-Gly-OH
SDP
Supplier:  Bachem 4002969.1000
Encompass_Preferred
H-Cys-Gly-OH, 19246-18-5, C5H10N2O3S, Mr 178.21, 1g. This dipeptide found in plasma and urine is generated during the catabolism of glutathione. Cysteinylglycine has been suggested to induce oxidative stress and lipid peroxidation, leading to the development of human cancers.
Catalog No. 50-259-417