Learn More
Medchemexpress LLC H-Cys(Z)-OH | 1625-72-5 | MFCD00055774 | 99.6% | 255.29 g/mol | C11H13NO4S | 25 G

Supplier: Medchemexpress LLC 25GHYW041997
H-Cys(Z)-OH is the benzyloxycarbonyl (Z) protected derivative of the amino acid cysteine, supplied as a solid reagent for research and peptide synthesis applications.
- Benzyloxycarbonyl-protected cysteine derivative used as a protected building block in peptide synthesis.
- High purity: 99.6%.
- Chemical formula C11H13NO4S; molecular weight 255.29 g/mol.
- Available in multiple pack sizes including 25 G.
- Documentation available: SDS, data sheet, and certificate of analysis (COA).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.