Learn More
Apexbio Technology LLC H-Cys(Trt)-OH 2799-07-7 25g
SDP
Supplier:  Apexbio Technology LLC A714325000
Encompass_Preferred
H-Cys(Trt)-OH (CAS 2799-07-7) also known as S-Trityl-L-cysteine is a small molecule allosteric inhibitor specifically targeting the kinesin motor protein Eg5 It exerts its effect by suppressing the ATPase activity of Eg5 both in its basal state and when activated by microtubule binding Eg5 is essential for mitotic spindle assembly and function inhibition of this protein disrupts mitosis and can impede the proliferation of tumor cells Consequently S-Trityl-L-cysteine is widely utilized in studies of cell cycle regulation and anti-tumor mechanisms
Catalog No. 50-224-0542