Learn More
Medchemexpress LLC H-D-Aha-OH hydrochloride | 1858224-26-6 | C4H9ClN4O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15172350MG
Encompass_Preferred
H-D-Aha-OH (hydrochloride) is a click chemistry reagent that contains an azide group. It can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have alkyne groups, and also in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Solid appearance
  • White to off-white color
  • Purity ≥98.0%
  • Suitable for click chemistry applications
  • Functions as an azide reagent
  • Useful as an ADC linker
  • Relevant for antibody-drug conjugate research
Catalog No. 50-003-75009