Learn More
Medchemexpress LLC H-D-alanine O-tert-butyl ester hydrochloride | 59531-86-1 | MFCD00077087 | >98.0% | 181.66 g·mol⁻¹ | C7H16ClNO2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01642610G
Encompass_Preferred
H-D-Ala-OtBu·HCl (D-alanine O-tert-butyl ester hydrochloride) is a protected D-alanine derivative used as a synthetic building block in peptide synthesis and medicinal chemistry research. It is supplied as a white to off-white solid with high purity and defined storage recommendations for stability.
  • High purity (≥98.0%) suitable for analytical and synthetic applications.
  • White to off-white solid form for easy handling and weighing.
  • Commonly used as a protected amino acid in peptide coupling and synthesis workflows.
  • Stable under recommended storage conditions to preserve quality.
  • Molecular weight and formula documented for formulation and calculation.
Catalog No. 50-003-02243