Learn More
Medchemexpress LLC H-D-Asp(OtBu)-OtBu·HCl | 135904-71-1 | >97.0% | 281.78 | 25 G

Supplier: Medchemexpress LLC 25GHYW013291A
H-D-Asp(OtBu)-OtBu·HCl is an isomer of H-Asp(OtBu)-OtBu.HCl and can be used as an experimental control. It is an aspartic acid derivative.
- Isomer of H-Asp(OtBu)-OtBu.HCl
- Can be used as an experimental control
- Aspartic acid derivative
- Appearance: solid
- Color: white to off-white
- Shipping: room temperature in continental US; may vary elsewhere
- Storage: 4°C, sealed storage, away from moisture
- Solubility in DMSO: 200 mg/mL (709.77 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.