Learn More
Medchemexpress LLC H-D-CHA-Ala-Arg-pNA diacetate | 99.83% | 638.71 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP6027A1MG
Encompass_Preferred
H-D-CHA-Ala-Arg-pNA diacetate is the diacetate form of H-D-CHA-Ala-Arg-pNA. It serves as a chromogenic substrate for the amidolytic assay.
  • Functions as a chromogenic substrate
  • Targets fluorescent dye applications
  • Belongs to the peptide classification
  • Useful as an enzyme substrate
Catalog No. 50-003-96355