Learn More
Medchemexpress LLC H-D-Chg-OH hydrochloride | 61367-40-6 | ≥98.0% | 193.67 | C8H16ClNO2 | 5 G

Supplier: Medchemexpress LLC HYW0507855G
H-D-Chg-OH hydrochloride is an amino acid derivative (a glycine derivative) supplied as the hydrochloride salt for research use. It is a white to off-white crystalline powder with high purity, intended for use as a reagent in peptide synthesis and biochemical research.
- High purity ≥98.0%
- Cas number 61367-40-6
- Molecular formula C8H16ClNO2
- Molecular weight 193.67
- Available in small pack sizes for research, including 5 G
- Powder storage: -20°C for long-term stability
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.