Learn More
Apexbio Technology LLC H-D-Dab-OH•2HCl 127531-11-7 25g
SDP
Supplier:  Apexbio Technology LLC A749325000
Encompass_Preferred
H-D-Dab-OH 2HCl is a synthetic amino acid derivative structurally characterized by a D-configured 2 4-diaminobutyric acid framework It serves as a building component for peptide synthesis enabling modification of peptide drug characteristics such as metabolic stability receptor binding dynamics and bioactivity profiles H-D-Dab-OH 2HCl does not have a specified target or defined mechanism of action There are no reported data on its activity type experimental model or key pharmacological metrics Based on its properties H-D-Dab-OH 2HCl is primarily applied in the preparation of peptide-based therapeutics and pharmacological tools supporting investigations into protein structure activity relationships and receptor interaction mechanisms
Catalog No. 50-000-97139