Learn More
Medchemexpress LLC H-D-Phe(4-CN)-OH | 263396-44-7 | MFCD00270364 | 98.0% | 190.20 g/mol | C10H10N2O2 | 5 G

Supplier: Medchemexpress LLC HYW0420065G
H-D-Phe(4-CN)-OH is a D-phenylalanine derivative bearing a 4-cyano substituent on the aromatic ring. It is provided for research use, commonly employed in peptide synthesis and studies of amino acid analogs.
- Phenylalanine derivative with 4-cyano substitution
- Suitable for peptide synthesis and amino acid research
- High purity (98.0%)
- Chemical formula C10H10N2O2; molecular weight 190.20 g/mol
- Available in laboratory-scale quantities (5 g, 10 g, 25 g, 100 g)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.