Learn More
TARGETMOL CHEMICALS INC H-D-Phe-Pip-Arg-pNA dihydro
SDP
Supplier:  TARGETMOL CHEMICALS INC T407111MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. H-D-Phe-Pip-Arg-pNA dihydrochloride (S-2238 dihydrochloride), a chromogenic substrate, mimics the N-terminal fragment of the A alpha chain of fibrinogen, the native substrate of thrombin. Specifically designed for thrombin detection, it is employed for quantifying antithrombin-heparin cofactor (AT-III) and is characterized by its sensitivity, accuracy, and ease of implementation. Purity 99.47%
Catalog No. 50-287-7162