Learn More
Medchemexpress LLC H-D-Phg(4-Cl)-OH | 43189-37-3 | MFCD03839815 | 97.0% | 185.61 g/mol | C8H8ClNO2 | 25 G

Supplier: Medchemexpress LLC 25GHYW042008
H-D-Phg(4-Cl)-OH is a chlorinated phenylglycine derivative (CAS 43189-37-3) used as a research reagent and amino acid derivative. It is supplied at research-grade purity for laboratory studies and peptide synthesis; not for human or clinical use.
- High purity (97.0%).
- Molecular weight 185.61 g/mol.
- Chemical formula C8H8ClNO2.
- Available in a 25 g package.
- Intended for research use only.
- Suitable as a building block in peptide synthesis and biochemical research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.