Learn More
MSC Value Collection Ball Transfer, Chrome Steel, Square Base, 1 ea.
Supplier: MSC 00290908
This ball transfer unit features a durable chrome steel ball housed in a sturdy steel frame with a square base, designed for efficient material handling and transport applications.
- Ball material: chrome steel
- Housing material: steel
- Base shape: square
- Mount height: 2-3/8 in.
- Overall diameter: 2-3/8 in.
- Load capacity: 2,425 lb.