Learn More
Medchemexpress LLC H-D-Tyr(Propargyl)-OH | 1170674-20-0 | C12H13NO3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15165350MG
Encompass_Preferred
H-D-Tyr(Propargyl)-OH is a click chemistry reagent containing an alkyne group. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing azide groups, making it suitable for various bioconjugation applications. This product is intended for research use only.
  • Alkyne group for click chemistry applications
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Suitable for antibody-drug conjugate (ADC) related research
  • Solid appearance, light yellow to yellow color
  • Purity of 95.0%
Catalog No. 50-003-73392