Learn More
Medchemexpress LLC H-Dab(Boc)-OMe hydrochloride | 3350-15-0 | 268.74 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW005561500MG
Encompass_Preferred
H-Dab(Boc)-OMe hydrochloride is an N-terminally protected diaminobutyric acid containing two protecting groups: methoxy (OMe) and tert-butyloxycarbonyl (Boc). It can be used to synthesize the bifunctional chelator H3Dpaa that can rapidly complex 68Ga under physiological conditions.
  • N-terminally protected diaminobutyric acid
  • Contains two protecting groups: methoxy (OMe) and tert-butyloxycarbonyl (Boc)
  • Synthesizes the bifunctional chelator H3Dpaa
  • Rapidly complexes 68Ga under physiological conditions
Catalog No. 50-004-47350