Learn More
Apexbio Technology LLC H-DL-Nva-OH 760-78-1 25g
SDP
Supplier:  Apexbio Technology LLC A765625000
Encompass_Preferred
H-DL-Nva-OH (CAS 760-78-1) also known as DL-Norvaline is the racemic form of norvaline structurally analogous to arginine It acts as a non-competitive inhibitor of arginase an enzyme involved in the conversion of arginine to ornithine and urea By inhibiting arginase activity DL-Norvaline modulates arginine metabolism and influences the biosynthesis of nitric oxide This compound is widely used in cellular and disease models to investigate pathways related to arginine metabolism particularly in studies of nitric oxide regulation and immunometabolic research
Catalog No. 50-000-93562