Learn More
Medchemexpress LLC H-DL-Phe(4-Me)-OH | 4599-47-7 | MFCD05227938 | 99.7% | 179.22 g/mol | C10H13NO2 | 5 G

Supplier: Medchemexpress LLC HYW0420015G
H-DL-Phe(4-Me)-OH is a phenylalanine derivative provided as a research-grade amino acid derivative for use as a building block or analytical standard in biochemical and medicinal chemistry research. It is supplied with documentation to support laboratory use.
- High purity (99.7%).
- Molecular weight 179.22 g/mol.
- Molecular formula C10H13NO2.
- Available in small laboratory quantities, including 5 G.
- Analytical documentation provided: COA and SDS.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.