Learn More
Medchemexpress LLC H-DL-Phe(4-NO2)-OH | 2922-40-9 | MFCD00007384 | 99.6% | 210.19 g/mol | C9H10N2O4 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW014405100G
Encompass_Preferred
H-DL-Phe(4-NO2)-OH is a para-nitro substituted DL-phenylalanine used as an amino acid derivative and synthetic building block in chemical and biochemical research. Supplied as a high-purity crystalline solid, it is suitable for peptide synthesis, method development, and as a reference standard for analytical and spectroscopic studies.
  • High purity (99.6%) solid suitable for research use.
  • Para-nitro substitution alters electronic and spectroscopic properties.
  • Useful as a building block in peptide synthesis.
  • Appropriate as a reference standard for analytical methods.
  • Compatible with common organic solvents and aqueous buffers.
  • Provided as a DL mixture (racemate) for non-chiral applications.
Catalog No. 50-003-02861