Learn More
Medchemexpress LLC H-dl-phe-otbu.hcl | 75898-47-4 | MFCD00190980 | > 99.0% | 257.76 g·mol⁻¹ | C13H20ClNO2 | 100 G

Supplier: Medchemexpress LLC HYW052408100G
H-DL-Phe-OtBu·HCl is a protected phenylalanine derivative supplied as the hydrochloride salt, used as an amino acid building block and intermediate in peptide synthesis and related research applications.
- Used as an amino acid building block in peptide synthesis
- Typical purity ≥ 99.0% (HPLC)
- Molecular weight 257.76 g·mol⁻¹
- Formula C13H20ClNO2
- Solid form suitable for storage as powder or in solvent
- Storage (powder): -20°C (3 years), 4°C (2 years)
- Storage (in solvent): -80°C (6 months), -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.