Learn More
Genscript Corporation h-FABP 27C1C mAb Mouse
Supplier:  Genscript Corporation V00901100
Encompass
Heart-type fatty acid binding protein (H-FABP) is encoded by the FABP3 gene with a low molecular weight of 15 kDa It is released in large quantities into blood at an early stage of acute myocardial infarction (AMI) H-FABP is a serum marker with high sensitivity and specificity for diagnosing AMI and myocardial ischemia
Catalog No. 50-000-58407
Only null left
Add to Cart
Request a Quote