Learn More
Medchemexpress LLC H-γ-Glu-Leu-OH | 2566-39-4 | 99.2% | 260.29 | 5 MG

Supplier: Medchemexpress LLC HYP46325MG
H-γ-Glu-Leu-OH (γ-Glu-Leu) is a dipeptide composed of γ-glutamic acid and leucine, featuring a hydroxyl group termination. It is suitable for various research applications.
- Dipeptide consisting of γ-glutamic acid and leucine
- Features a hydroxyl group termination
- Purity of 99.2%
- Molecular weight: 260.29
- Soluble in DMSO (≥ 100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.