Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Glu(OMe)-OH | 1499-55-4 | 98.0% | 161.16 | 100 G
SDP

Supplier:  Medchemexpress LLC HYW013678100G

Encompass_Preferred

H-Glu(OMe)-OH is a glutamic acid derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements, influencing the secretion of anabolic hormones, fuel supply during exercise, mental performance during stress-related tasks, and preventing exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.

  • Solid appearance
  • White to off-white color
  • Ships at room temperature in continental US; conditions may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-54500


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.