Learn More
Medchemexpress LLC H-Gly-Ala-Hyp-OH | 22028-81-5 | 99.6% | 259.26 | 5 MG

Supplier: Medchemexpress LLC HYP42315MG
H-Gly-Ala-Hyp-OH is a collagen tripeptide fragment.
- For research use only
- Solid, white to off-white appearance
- Sequence: H-Gly-Ala-Hyp-OH
- Exhibits a Km value of 2.080 mM
- Soluble in DMSO up to 100 mg/mL (385.71 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.