Learn More
Bachem H-Gly-Gly-His-OH
SDP
Supplier:  Bachem 4006117.1000
Encompass_Preferred
H-Gly-Gly-His-OH, 7451-76-5, C10H15N5O4, Mr 269.26, 1g. Cu(II) and Ni(II)-binding tripeptide.The Ni(II) complex of GGH was shown by Brown et al. to mediate protein crosslinking by oxidants. GGH has been used in biosensors for the electrochemical detection of Cu(II) ions.
Catalog No. 50-259-748