Learn More
Medchemexpress LLC H-Gly-Gly-Pro-OH | 14379-76-1 | ≥99.0% | 229.24 | 1 G

Supplier: Medchemexpress LLC HY1119221G
H-Gly-Gly-Pro-OH is a tripeptide also known as Glycyl-glycyl-L-proline. This compound has a molecular weight of 229.24 and a CAS number of 14379-76-1. It is supplied as a white to off-white solid with a purity of ≥99.0%.
- Composed of three amino acids
- Molecular weight: 229.24
- Purity: ≥99.0%
- CAS number: 14379-76-1
- Appears as a white to off-white solid
- Soluble in water at ≥100 mg/mL
- Store as powder at -80°C for up to 2 years, or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.