Learn More
Medchemexpress LLC H-Gly-Leu-Phe-OH | 103213-38-3 | 99.4% | 335.40 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP428610MG
Encompass_Preferred
H-Gly-Leu-Phe-OH (GLF) is an immunostimulatory tripeptide derived from α-lactalbumin. It has been shown to inhibit alopecia, epidermal thickening, and the thinning of the adipocyte layer induced by the anticancer agent Etoposide. This makes it a potential subject for research in various biological contexts, especially concerning its protective effects against drug-induced side effects.
  • Immunostimulatory tripeptide.
  • Derived from α-lactalbumin.
  • Inhibits Etoposide-induced alopecia.
  • Prevents epidermal thickening and adipocyte layer thinning.
  • Suitable for research use.
Catalog No. 50-003-99735