Learn More
Bachem H-Gly-Pro-Arg-Pro-NH2
SDP
Supplier:  Bachem 4025347.0005
Encompass_Preferred
H-Gly-Pro-Arg-Pro-NH2 Acetate, 126047-75-4, C18H32N8O4, Mr 424.50, 5mg. The inhibitory effect of GPRP amide on fibrinogen/thrombin clotting is 3.52 times higher than that of the corresponding tetrapeptide GPRP.
Catalog No. 50-382-505