Learn More
Bachem H-Gly-Pro-Arg-Pro-OH
SDP
Supplier:  Bachem 4004545.0100
Encompass_Preferred
H-Gly-Pro-Arg-Pro-OH Acetate, 67869-62-9, C18H31N7O5, Mr 425.49, 100mg. Fibrinolysis Inhibiting Factor. GPRP (H-2935) inhibits fibrinogen/thrombin clotting, though it is less active than its C-terminal amide GPRP-NH2.
Catalog No. 50-259-668