Learn More
Medchemexpress LLC HY-W016887 100mg Medchemexpress, H-Gly-Pro-OH CAS:704-15-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW016887100MG
Encompass_Preferred
Medchemexpress, HY-W016887 100mg H-Gly-Pro-OH CAS:704-15-4 H-Gly-Pro-OH is an end product of collagen metabolism that is further cleaved by prolidase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0724