Learn More
Bachem H-Gly-Pro-pNA · HCl
SDP
Supplier:  Bachem 4025614.0250
Encompass_Preferred
H-Gly-Pro-pNA · HCl, 103213-34-9, C13H16N4O4 · HCl, Mr 328.76, 250mg. Excellent substrate for dipeptidylpeptidase IV (DPP IV).
Catalog No. NC1612287