Learn More
Medchemexpress LLC H-His(Trt)-OH | 35146-32-8 | 99.90% | 397.47 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW013155500G
Encompass_Preferred
H-His(Trt)-OH is a histidine derivative. Amino acids and amino acid derivatives, such as H-His(Trt)-OH, are commercially utilized as ergogenic supplements. They are known to influence the secretion of anabolic hormones, provide fuel during exercise, enhance mental performance in stress-related tasks, and help prevent exercise-induced muscle damage. These substances are recognized as beneficial ergogenic dietary components.
  • Histidine derivative
  • Ergogenic supplement
  • Influences anabolic hormones
  • Fuel during exercise
  • Enhances mental performance
  • Prevents muscle damage
  • Beneficial dietary substance
Catalog No. 50-004-54342