Learn More
Medchemexpress LLC H-His(trt)-OMe hydrochloride | 32946-56-8 | MFCD00237100 | 97.0% | 447.96 g·mol⁻¹ | C26H26ClN3O2 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0222201G
Encompass_Preferred
H-His(trt)-OMe hydrochloride is the trityl-protected L-histidine methyl ester hydrochloride used as a protected amino acid building block in peptide and small-molecule synthesis. The trityl (Trt) group shields the imidazole side chain during coupling and deprotection, while the methyl ester and hydrochloride salt forms aid handling, storage, and downstream transformations.
  • Protects histidine imidazole side chain during synthesis.
  • Compatible with solid-phase and solution-phase peptide synthesis.
  • Methyl ester facilitates coupling and controlled deprotection.
  • Hydrochloride salt improves stability and handling.
  • High purity suitable for synthetic applications.
Catalog No. 50-003-00966